Order Entry
United States
ContactUsLinkComponent
 
Anti-CD8A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: Hit8a]
 
undefined
Anti-CD8A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: Hit8a]
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Catalog No:
  • 10208-408
  • 10208-406
  • Antigen Symbol:
  • CD8A
  • CD8A
  • Antigen Name:
  • CD8 antigen, alpha chain
  • CD8 antigen, alpha chain
  • Antigen Synonyms:
  • p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
  • p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • Hit8a
  • Hit8a
  • Reactivity:
  • Rhesus monkey, Chimpanzee, Human
  • Rhesus monkey, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 925
  • 925
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Storage Temperature:
  • 2-8°C protected from light
  • 2-8°C protected from light
  • Tested Applications:
  • Flow Cytometry
  • Flow Cytometry