ContactUsLinkComponent
Solvent Extractors According to Randall, VELP Scientifica
Solvent Extractors According to Randall, VELP Scientifica
Certificates
undefined
Solvent Extractors According to Randall, VELP Scientifica
Specifications
  • Catalog No:
  • 10184-156
  • 10184-158
  • Model:
  • SER 148/3
  • SER 148/6
  • No. of samples:
  • 3
  • 6
  • Sample capacity:
  • 0.5 to 15 g (typically 2 - 3 g)
  • 0.5 to 15 g (typically 2 - 3 g)
  • Temperature Operating Range:
  • 100 - 260 °C
  • 100 - 260 °C
  • Displayed results:
  • Working temperature/immersion time/washing time/recovery time
  • Working temperature/immersion time/washing time/recovery time
  • Immersion Time:
  • 0 - 999 min
  • 0 - 999 min
  • Washing Time:
  • 0 - 999 min
  • 0 - 999 min
  • Solvent Recovery (typical):
  • 50 - 75%
  • 50 - 75%
  • Water Consumption (l):
  • 120 l/h
  • 120 l/h
  • No. of programs:
  • 29
  • 29
  • Housing:
  • Epoxy painted stainless steel structure
  • Epoxy painted stainless steel structure
  • Max. Volume of Extraction Cup:
  • 150
  • 150
  • Recovery Time:
  • 0 - 999 min
  • 0 - 999 min
Specifications