Order Entry
Puerto Rico
ContactUsLinkComponent
Bromophenol blue, powder
Bromophenol blue, powder
Catalog # IC805732
Supplier:  MP Biomedicals
CAS Number:  
Bromophenol blue, powder
Catalog # IC805732
Supplier:  MP Biomedicals
Supplier Number:  04805732
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    10 g
  • Cat. No.:
    IC805732
  • Supplier no.:
    805732

Specifications

About this item

 :
 :669.97 g/mol
Melting Pt:273 °C (lit.)
EINECS:204-086-2
CAS Number:115-39-9
Formula:C₁₉H₁₀Br₄O₅S
MDL Number:MFCD00005875