Glutaraldehyde 25% in aqueous solution
Catalog # IC19519910
Supplier: MP Biomedicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:10 ml
- Cat. No.:IC19519910
- Supplier no.:19519910
Specifications
Specification Test Results
- Synonyms:Glutaric dialdehyde; 1,3-Diformyl propane; Cidex; Glutarol-1,5-pentanedial; 1,5-Pentanedione
- Form:Clear Liquid
- Storage:Store at +4 °C, store under nitrogen.
- pH:3.1 - 4.5
- Viscosity:8.4 cps (0°C), 3.4 cps (20°C), 1.7 cps (40°C) (Lit.)
Specification Test Results
About this item
:
:100.12
UN:2927
ADR:6.1,II
CAS Number:111-30-8
Formula:C₅H₈O₂
MDL Number:MFCD00007025