Order Entry
Puerto Rico
ContactUsLinkComponent
Collagenase, Type I (from Clostridium histolyticum), MP Biomedicals
Collagenase, Type I (from Clostridium histolyticum), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Collagenase, Type I (from Clostridium histolyticum), MP Biomedicals
Supplier:  MP Biomedicals
Certificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • CAS No.:
    9001-12-1
  • Source:
    Clostridium histolyticum
  • Storage Conditions:
    Store at +4 °C, desiccate
  • Form:
    Brown lyophilized powder
  • Enzyme Name:
    Collagenase, Type I
  • Solubility:
    Dissolve enzyme at a concentration of 1 mg/ml in 0.05 M TES with 0.36 mM calcium chloride, pH 7.5.
  • MDL No.:
    MFCD00130830

Specifications

Related Information

Frequently Bought Together