Order Entry
Puerto Rico
ContactUsLinkComponent
Pyruvate Oxidase, MP Biomedicals
Pyruvate Oxidase, MP Biomedicals
Catalog # IC15348625
Supplier:  MP Biomedicals
CAS Number:  
Pyruvate Oxidase, MP Biomedicals
Catalog # IC15348625
Supplier:  MP Biomedicals
Supplier Number:  0215348625
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    25 U
  • Form:
    Powder
  • Enzyme Name:
    Pyruvate Oxidase
  • Cat. No.:
    IC15348625
  • Supplier no.:
    15348625

Specifications

About this item

One unit causes the formation of one µmole of hydrogen peroxide (half a µmole of quinoneimine dye) per minute at pH 5.7 at 37 °C.

Michaelis constant : 3.4 X 10^-4M (Pyruvate) Inhibitors : Fe++,Zn++,Cu++,Ag+,Hg++ Optimum temperature : 65°C, pH Stability : pH 5.7 - 6.5 (25°C,, 20hr) Thermal stability : below 45°C, (pH 6.0, 15min)

Pyruvate Oxidase reacts with certain aldehydes and phosphate can be replaced by arsenate. Oxygen as well as several artificial compounds can function as electron acceptors. Pyruvate Oxidase is activated by phospholipids as well as monomeric and micellar amphiphiles.