Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- CAS No.:17650-98-5
- Size:1 mg
- Storage Conditions:Freezer
- Form:White powder
- MDL No.:MFCD00076478
- Protein Synonyms:pyroGlu-Gln-Asp-Tyr[SO3H]-Thr- Gly-Trp-Met-Asp-Phe-NH2
- Protein/Peptide Name:Caerulein, sulfated
- Molecular Weight:1352.412 g/mol
- Molecular Formula:C₅₈H₇₃N₁₃O₂₁S₂
- Cat. No.:IC15286001
- Supplier no.:15286001
Caerulein is a gastroregulatory molecule with actions similar to cholecystokinin. Product supplied as a trifluoroacetate salt.
Caerulein is used to study cell signaling mediated by NF-kappaB upregulation of proteins such as intercellular adhesion molecule-1(ICAM-1) and inflammation related molecules such as NADPH oxidase and Janus kinase signal transducer. It prevents gall bladder pain, renal colic, and the pain of intermittent claudication.
It is seen as an antagonist of endogenous opioids.
Soluble in ammonium hydroxide 0,05 M (1 mg/ml)(Lit.)
Sequence: pGlu-Gln-Asp-Tyr(SO3H)-Thr-Gly-Trp-Met-Asp-Phe-NH2
Caerulein is used to study cell signaling mediated by NF-kappaB upregulation of proteins such as intercellular adhesion molecule-1(ICAM-1) and inflammation related molecules such as NADPH oxidase and Janus kinase signal transducer. It prevents gall bladder pain, renal colic, and the pain of intermittent claudication.