Specifications
- Size:10 KU
- Form:Powder
- Enzyme Name:Macrozyme
- Cat. No.:IC15234010
- Supplier no.:15234010
Specifications
About this item
Readily soluble in water.
The amount of macerating activity on potato tuber disks as determined by weight loss at 40 °C, pH 5.0.