Order Entry
Puerto Rico
ContactUsLinkComponent
 
Red Kidney Beans (Phaseolus vulgaris) Phytohemagglutinin, MP Biomedicals
Catalog # IC15188601
Supplier:  MP Biomedicals
CAS Number:  
Red Kidney Beans (Phaseolus vulgaris) Phytohemagglutinin, MP Biomedicals
Catalog # IC15188601
Supplier:  MP Biomedicals
Supplier Number:  0215188601
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Species:
    Red kidney beans (Phaseolus vulgaris)
  • Size:
    1 mg
  • Storage Conditions:
    Store at -0 °C. Store solutions sterile, filtered and frozen in aliquots. Avoid freeze-thaw cycles. Clarify by centrifugation.
  • Form:
    White Solid
  • Protein Synonyms:
    PHA-P|PHA|Phaseolus vulgaris agglutinin|PHA-L|Lectin from Phaseolus vulgaris
  • Protein/Peptide Name:
    Phytohemagglutinin
  • Cat. No.:
    IC15188601
  • Supplier no.:
    15188601

Specifications

About this item

Reconstitution: Reconstitute with water to a final concentration of 1 mg/mL (use 1 mL for the 1 mg size or 5 mL for the 5 mg size) for a final solution of 1 mg/mL in 0.40 mM sodium phosphate, 6 mM NaCl, pH 7.45. Reconstitute with water to a final concentration of 25 mg/mL (0.2 ml water for the 5 mg size). This concentration of the lectin has been used for iontophoresis and neuron transport.

Carbohydrate Specificity: PHA-L recognizes a complex branched chain oligosaccharide structure.