Order Entry
Puerto Rico
ContactUsLinkComponent
2'-Deoxyadenosine-5'-diphosphate trisodium salt ≥97%, white powder
Toxic
2'-Deoxyadenosine-5'-diphosphate trisodium salt ≥97%, white powder
Catalog # IC15079410
Supplier:  MP Biomedicals
CAS Number:  
Toxic
2'-Deoxyadenosine-5'-diphosphate trisodium salt ≥97%, white powder
Catalog # IC15079410
Supplier:  MP Biomedicals
Supplier Number:  0215079410
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    10 mg
  • Cat. No.:
    IC15079410
  • Supplier no.:
    15079410

Specifications

Specification Test Results

  • Identity Test :
    Passes
  • Purity:
    ≥97%

Specification Test Results

About this item

 :
 :455.167 g/mol
 :GHS06
Storage Temperature:Freezer
CAS Number:72003-83-9
Formula:C₁₀H₁₃N₅Na₂O₉P₂
MDL Number:MFCD00083610