About this item
Pure Canavalia ensiformis lectin (Con A) from Jackbean. Isolated by affinity chromatography on cross-linked dextran. Con A exists as a dimer below pH 5.0 and and at pH >7 it exists as a tetramer. Con-A is not a glycoprotein. The monomeric molecular weight of Con-A is 25500. Con-A does not contain cysteine residues. Unlike most other lectins, Con-A is a metalloprotein and requires a transition metal ion, such as manganese, plus calcium ions for binding.
- Isoelectric point (pl): 4.5, 4.7, 5.0 to 5.1, and 5.4 to 5.5 (several isoforms)(Lit.)
- Inhibitory Carbohydrate: Methyl-a-D-Mannopyranoside >> a-D-Mannose >> a-D-Glucose
- Carbohydrate Specificity: a-Mannose, a-Glucose, Branched Mannose
- Soluble in water (10 mg/ ml-slightly hazy, colorless solution)
- GHS symbols and signal word: GHS07, GHS08 and Danger
Lectin are used in a wide variety of applications in vitro, including: blood grouping and erythrocyte polyagglutination studies, Mitogenic stimulation of lymphocytes, Lymphocyte subpopulation studies, Fractionation of cells and other particles, Histochemical studies of normal and pathological conditions. Con-A has been used to elucidate structural changes in the membrane surface of transformed cells. Con-A conjugates of antitumor drugs have been used in drug delivery systems in cultured cells.
Con A is not blood group specific but has an affinity for terminal α-D-mannosyl and α-D-glucosyl residues. Ca²⁺ and Mn²⁺ ions are required for activity. Con A dissociates into dimers at pH 5.6 or below. Between pH 5.8 and pH 7.0, Con A exists as a tetramer; above pH 7.0 higher aggregates are formed. Con A exhibits mitogenic activity which is dependent on its degree of aggregation. Succinylation results in an active dimeric form which remains a dimer above pH 5.6. Con-A binds specifically to mannosyl and glucosyl residues of polysaccharides and glycoproteins. Unmodified hydroxyl groups at the C3, C4 and C6 positions of D-glucopyranosyl or D-mannopyranosyl rings may be essential for binding. Each subunit of Con-A contains one calcium ion and one manganese ion. Removal of these cations by dialysis under acidic conditions abolishes the carbohydrate-binding activity.
Inhibitory Carbohydrate: Methyl-a-D-Mannopyranoside a-D-Mannose a-D-Glucose.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:11028-71-0
- Source:Jack beans
- Storage Conditions:–20 °C
- Form:White Shiny Powder
- MDL No.:MFCD00071069
- Protein/Peptide Name:Concanavalin A