2-Phenoxyethanol
About this item
Important Notice: This product belongs to MP Biomedicals Rare Chemical library. Most Rare Chemical products are available for immediate delivery and sold "As Is". MP Biomedicals does not collect analytical data for those products. All sales are final. However, analytical test may be performed upon custom request for bulk orders.
Caution: Research Use Only (RUO).
- CAS Number:122-99-6
- Formula:C₆H₅OCH₂CH₂OH
- MDL Number:MFCD00002857
- Molecular Weight:138.17 g/mol
- Melting Pt:57 ° F (lit.)
- Storage Temperature:Ambient
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Important Notice: This product belongs to MP Biomedicals Rare Chemical library. Most Rare Chemical products are available for immediate delivery and sold "As Is". MP Biomedicals does not collect analytical data for those products. All sales are final. However, analytical test may be performed upon custom request for bulk orders.
Caution: Research Use Only (RUO).
- CAS Number:122-99-6
- Formula:C₆H₅OCH₂CH₂OH
- MDL Number:MFCD00002857
- Molecular Weight:138.17 g/mol
- Melting Pt:57 ° F (lit.)
- Storage Temperature:Ambient
Documents
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications for Product Family
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: 0215014880Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: 0215014890Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



