Excess cholesterol is stored intracellularly as cholesterol esters.
- Presentation: Yellowish tan lyophilized powder
- Isoelectric point (pl): 5.9 ±0.1(Lit.)
- Optimum pH: 7.0 to 9.0 ; pH stability: pH 5.0 to 9.0 (25 °C, 24 h)
- Soluble in distilled water or dilute buffer
Cholesterol Esterase is useful for enzymatic determination of total cholesterol when coupled with cholesterol oxidase in clinical analysis.
Cholesterol esterase (CE) is a reversible enzyme that can hydrolyze or synthesize fatty acid esters of cholesterol and other sterols. Hydrolysis of water insoluble long chain fatty acid esters requires bile salt activation. Hydrolysis of water soluble esters of short chain fatty acids and lysophospholipids does not require activation by bile salts. It also hydrolyzes tri-, di-, and mono-acylglycerols, phospholipids, lysophospholipids, and ceramide. The enzyme may have multiple functions in lipid and lipoprotein metabolism, and atherosclerosis.
Cholesterol esterase (CE) is a reversible enzyme that can hydrolyze or synthesize fatty acid esters of cholesterol and other sterols.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- CAS No.:9026-00-0
- Storage Conditions:–20 °C
- Form:Yellowish Tan Lyophilized Powder
- Enzyme Name:Cholesterol Esterase
- Enzyme Activity:≥100 U/mg solid