L-Leu-L-Ala, MP Biomedicals
Catalog # IC10216701
Supplier: MP Biomedicals
CAS Number:
Specifications
- Size:1 g
- Protein/Peptide Name:L-Leu-L-Ala
- Cat. No.:IC10216701
- Supplier no.:10216701
Specifications
About this item
Soluble in acetic acid 80% (50 mg/ml-clear colorless solution)
- Synonyms: Leu-Ala; H-Leu-Ala-OH
- Form: White Crystalline Powder
- Storage: 0 °C
- Optical Rotation: [α]20/D +19±3°, c = 2% in methanol
Used as fluorescence intensifier in fluorimetric methods for the determination of phenylalanine.