Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Dextran

Dextran

Dextran
Supplier:  MP Biomedicals
Certificates

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Dextran is a high molecular weight polymer of anhydroglucose. It is composed of approximately 95% alpha-D-(1-->6) linkages. Dextrans with molecular weights greater than 10,000 behave as if they are highly branched. As the molecular weight increases, dextran molecules attain greater symmetry. Solutions of 10% dextran (MW ~40,000) exert a slightly higher colloidal osmotic pressure than plasma proteins.

Dextran solutions have been used as plasma extenders. Colloidal solutions containing dextran MW 250,000 have been used to isolate aggregated platelets leukocytes and lymphocytes in blood by centrifugation. Dextran MW 40,000 has been used for the isolation of intact nuclei. Increase in threonine, serine, proline, and cysteine dietary supply can promote mucin synthesis in dextran sulfate sodium treated rats. Dextran (MW ~400,000 to 500,000) can be used as long and hydrophilic spacer arms to improve the performance of immobilized proteins acting on macromolecules. Dextran 40 is used in the development of new improved preservation solutions for organ transplantation. Dextran 40 may be used to determine cell tightness and flux parameters across cell layers.

Soluble in water (> 30 mg/mL), freely soluble in DMSO, formamide, ethylene glycol and glycerol. Neutral aqueous dextran solutions can be sterilized by autoclaving at 110-115 °C for 30 to 45 minutes.

Dextran is a high molecular weight polymer of anhydroglucose. It is composed of approximately 95% alpha-D-(1--6) linkages. Dextrans with molecular weights greater than 10,000 behave as if they are highly branched. As the molecular weight increases, dextran molecules attain greater symmetry. Solutions of 10% dextran (MW ~40,000) exert a slightly higher colloidal osmotic pressure than plasma proteins.

  • CAS Number:9004-54-0
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00130935
Specifications for Product Family

Specification Test Results

  • Form:
    White Powder
  • Storage:
    Store at Room Temperature (15-30 °C).
  • Loss on Drying:
    ≤5.0%
  • Optical Rotation:
    +192 ° to +204 °
  • pH:
    5.0 - 7.0
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 10 g
    Log in for availability
    Log in for availability
    Each (10g)
    Each (10g) - $56.20
    $56.20
    Each (10g)
       $56.20

    Supplier Number: 0210150510
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 50 g
    Log in for availability
    Log in for availability
    Each (50g)
    Each (50g) - $259.00
    $259.00
    Each (50g)
       $259.00

    Supplier Number: 0210150550
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $348.00
    $348.00
    Each (100g)
       $348.00

    Supplier Number: 0210150580
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.