Caffeine monohydrate, white powder
About this item
Caffeine Monohydrate protects against virus induction and chemical carcinogenesis. Commonly used as a mild stimulant.
It overrides the cell cycle effects of various chemicals such as protease inhibitors, thereby preventing apoptosis and it has been shown to inhibit cellular DNA repair mechanisms.
A central nervous system stimulant believed to act through adenosine receptors and monoamine neurotransmitters. It is an adenosine receptor antagonist and adenosine 3′,5′-cyclic monophosphate (cAMP) phosphodiesterase inhibitor. Thus, levels of cAMP increase in cells following treatment with caffeine. It has been reported to affect cellular calcium levels, releasing calcium from intracellular stores. It overrides the cell cycle effects of various chemicals such as protease inhibitors, preventing apoptosis; and it has been shown to inhibit cellular DNA repair mechanisms.
Soluble in hot water.
Caffeine Monohydrate protects against virus induction and chemical carcinogenesis. commonly used as a mild stimulant. It overrides the cell cycle effects of various chemicals such as protease inhibitors, thereby preventing apoptosis and it has been shown to inhibit cellular DNA repair mechanisms.
- CAS Number:58-08-2
- Formula:C₈H₁₀N₄O₂·H₂O
- MDL Number:MFCD00005758
- Molecular Weight:212.2 g/mol
- Melting Pt:230…240 °C
- Density:1.23 g/cm³ (lit.)
- Storage Temperature:Ambient
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Caffeine Monohydrate protects against virus induction and chemical carcinogenesis. Commonly used as a mild stimulant.
It overrides the cell cycle effects of various chemicals such as protease inhibitors, thereby preventing apoptosis and it has been shown to inhibit cellular DNA repair mechanisms.
A central nervous system stimulant believed to act through adenosine receptors and monoamine neurotransmitters. It is an adenosine receptor antagonist and adenosine 3′,5′-cyclic monophosphate (cAMP) phosphodiesterase inhibitor. Thus, levels of cAMP increase in cells following treatment with caffeine. It has been reported to affect cellular calcium levels, releasing calcium from intracellular stores. It overrides the cell cycle effects of various chemicals such as protease inhibitors, preventing apoptosis; and it has been shown to inhibit cellular DNA repair mechanisms.
Soluble in hot water.
Caffeine Monohydrate protects against virus induction and chemical carcinogenesis. commonly used as a mild stimulant. It overrides the cell cycle effects of various chemicals such as protease inhibitors, thereby preventing apoptosis and it has been shown to inhibit cellular DNA repair mechanisms.
- CAS Number:58-08-2
- Formula:C₈H₁₀N₄O₂·H₂O
- MDL Number:MFCD00005758
- Molecular Weight:212.2 g/mol
- Melting Pt:230…240 °C
- Density:1.23 g/cm³ (lit.)
- Storage Temperature:Ambient
Documents
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications for Product Family
Specification Test Results
- identity Test:Passes
- FTIR:Conforms to Standard
- Melting Point:230 - 240°C
- Moisture Content:4 - 8%
- pH:6 - 7.5 (1% aq soln)
- UV/Vis: l max (water):271 - 273 nm
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: 0210119480Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: 0210119491Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



