Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Bromothymol blue, powder ACS

Bromothymol blue, powder ACS

5 g
Catalog # IC10113005
Supplier:   MP Biomedicals
Supplier #:  0210113005
Bromothymol blue, powder ACS
Catalog # IC10113005
Supplier:  MP Biomedicals
CAS Number:  76-59-5
Bromothymol blue, powder ACS

5 g

CAS Number:  76-59-5
Price
$35.20
Log in for availability
Each (5g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Bromothymol Blue (BTB) is a pH indicator (pKa approx. 7.1) that is used as a biological slide vital stain to define cell walls or nuclei under the microscope. BTB is also a component of several bacterial growth and detection substrates such as cystine lactose electrolyte deficient (CLED/BROLACIN); polymyxin pyruvate egg yolk mannitol bromothymol blue (PEMBA) and MacConkey agar.

Soluble in water (Sparingly soluble), ethanol, ether, and aqueous solutions of alkalis.

Bromothymol Blue (BTB) is a pH indicator (pKa approx. 7.1) that is used as a biological slide vital stain to define cell walls or nuclei under the microscope. BTB is also a component of several bacterial growth and detection substrates such as cystine lactose electrolyte deficient (CLED/BROLACIN); polymyxin pyruvate egg yolk mannitol bromothymol blue (PEMBA) and MacConkey agar.

Synonyms:  3′,3″-Dibromothymolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:76-59-5
  • Formula:C₂₇H₂₈Br₂O₅S
  • MDL Number:MFCD00005872
  • Molecular Weight:624.39 g/mol

Documents

Specifications

Specifications

  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.