One unit will solublize 1 mg of elastin in 20 minutes at 37 °C, pH 8.8.
- Inhibitors: DFP, elastinal, α-2-macroglobulin
Elastase is prepared from porcine pancreas. It is lyophilized and water soluble. Elastase is chromatographically prepared by the method of Narayanan and Anwar. In the method, 2 times crystalline elastase is adsorbed on a column of DEAE-Sephadex A50 to separate elastase and non-specific protein components. The elastase component is further purified by chromatography on a column of CM-Cellulose using a sodium chloride gradient to elute the elastase. The latter is dialyzed until chloride-free and then lyophilized. During its preparation the elastase is held below a pH of 5.5 for greater than 24 hours. Two times crystallized from the euglobin fraction of porcine pancreas by the method of Lewis et al. Does not contain trypsin or chymotrypsin.
Elastase is an enzyme from the class of proteases (peptidases) that break down proteins. It hydrolyzes peptide bonds, especially those adjacent to neutral amino acids.
Elastase was used to study human emphysema by induction of emphysema in rodents. It has been used in a study to investigate the design, synthesis and evaluation of biomimetic affinity ligands for elastases. Elastase from porcine pancreas has also been used in a study to investigate the purification and partial characterization of the pancreatic proteolytic enzymes trypsin, chymotrypsin, and elastase. It hydrolyzes peptide bonds, especially those adjacent to neutral amino acids. Elastase hydrolyses elastin, the specific protein of elastic fibers, and digests hemoglobin, casein and fibrin.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- CAS No.:9004-06-2
- Form:Solution
- Enzyme Name:Elastase
- Purity:2X crystallized