About this item
One unit will hydrolyze 1 µmole of N-benzoyl-L-tyrosine ethyl ester per minute at pH 7.8 and 25 °C. Produced from 3× crystallized chymotrypsinogen.
Inhibitors: The enzyme is inhibited by heavy metals, the natural trypsin inhibitors to various degrees, an inhibitor from potato, and organophosphorus compounds. Also inhibited by AEBSF, α-1-antitrypsin, Aprotinin, DFP, PMSF, TPCK and α-2-Macroglobulin.
Chymotrypsin preferentially catalyzes the hydrolysis of peptide bonds involving L-isomers of tyrosine, phenylalanine, and tryptophan. It also readily acts upon amides and esters of susceptible amino acids. In addition to bonds involving aromatic amino acids, chymotrypsin catalyzes at a high rate the hydrolysis of bonds of leucyl, methionyl, asparaginyl, and glutamyl residues. a-Chymotrypsin is a protein consisting of 241 amino acid residues. The molecule has three peptide chains: an A chain of 13 residues, a B chain of 131 residues, and a C chain of 97 residues.
α-Chymotrypsin is used for treating pancreatic insufficiency and in traumatology.
A serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains (Tyr, Trp, Phe, Met, Leu) on the carboxyl end of the bond.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:9004-07-3
- Storage Conditions:Store at +4 °C.
- Form:White powder
- Enzyme Name:α-Chymotripsin
- Enzyme Synonyms:E.C. 3.4.21.1
- Purity:3X Crystallized
- Concentration:Protein: ≥80%