This α-amylase product is a liquefying diastase supplied as a dry, free-flowing powder having high uniformity and excellent storage stability.
- Presentation: Tan Powder
- pH: 5 to 7 (optimum around 6.1)
The general procedure for the enzymatic liquefaction of starch is the same, regardless of the nature of the end product. The degree of conversion is followed by measuring the viscosity. The ultimate viscosity is controlled by the level of enzyme and the time which elapses after gelatinization. The usual conditions are 0.03 - 0.06% a-Amylase based on weight of starch for 15 to 30 minutes. The starch is suspended in cold water with agitation sufficient to assure uniform dispersion. If vigorous agitation is provided, the dry solid enzyme may be added directly to the suspension. A safer practice, however, is to dissolve the dry enzyme in a little water before charging. The reaction mixture is then heated to the gelatinization temperature and held there (or slightly hotter) until the required viscosity is reached. Potato, tapioca, sago, arrowroot, and cereal starches other than those obtained from maize or rice are gelatinized at 70 - 75 °C. Maize and rice starches require a temperature of 80 °C. When the conversion is complete, the mixture is heated quickly to 100 °C and held 10 minutes to inactivate the enzyme. Additional heating at 100 °C is sometimes done to effect further reduction in viscosity by the action of heat alone.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- CAS No.:9000-90-2
- Source:Bacillus subtilis
- Storage Conditions:Store at +4 °C, desiccate
- Form:Tan Powder
- Enzyme Name:alpha-Amylase
- Enzyme Activity:≥160000 BAU/g