Fibroblast Growth Factor 10, human
Formula Weight: 19.3 kDa
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White
MDL No.: MFCD01867039
Applications: Human KGF2 shares 92% and 95% amino acid sequence identity with mouse and rat KGF2, respectively. It is most closely related to FGF7, FGF3 and FGF22. Recombinant KGF2 has been shown to be mitogenic for epithelial and epidermal cells but not fibroblasts.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Protein/Peptide Type:Recombinant
- Species:Human
- Storage Conditions:−30 °C to −10 °C
- MDL No.:MFCD01867039
- Protein/Peptide Name:Fibroblast Growth Factor 10
- Molecular Weight:19.3 kDa
- Color:White
- Formulation:Lyophilized powder