About this item
A substitute for glutamine in mammalian cell culture media; stable to heat-sterilization.
- Boiling point: 615 °C
- EINECS: 475-290-9
- Flash point: >110 °C(230 °F)
- Melting point: 222 °C
Purity: 99% CAS No.: 39537-23-0 Molecular Formula: C8H15N3O4 Formula Weight: 217.22 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: White Melting Point: 222° Boiling Point: 615° Solubility: Soluble in water MDL No.: MFCD00133046
Applications: A substitute for glutamine in mammalian cell culture media; stable to heat-sterilization
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:39537-23-0
- Storage Conditions:Ambient
- MDL No.:MFCD00133046
- Protein Synonyms:H-Ala-Gln-OH
- Protein/Peptide Name:L-Alanyl-L-glutamine
- Purity:99%
- Molecular Weight:217.23 g/mol
- Molecular Formula:C₈H₁₅N₃O₄