Phosphorus, plasma standard solution, Specpure®, P 10,000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Phosphorus, plasma standard solution, Specpure|r, P 10,000µg/ml
Molecular Formula: H3PO4, 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00133771
Specifications
- MDL No.:MFCD00133771