Molybdenum, Plasma Standard Solution, Specpure®, Mo 10 µg/ml, Thermo Scientific
AA45260-AE
:
- Concentration:10 µg/ml
- Matrix:2% HNO₃
- Size:100 ml
- MDL No.:MFCD00003465
- Cat. No.:AA45260-AE
- Element:Molybdenum
- Supplier no.:45260-AE
Molybdenum, plasma standard solution, Specpure|r, Mo 10µg/ml
Molecular Formula: Mo in 2% HNO3/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00003465