Order Entry
Puerto Rico
ContactUsLinkComponent
 
CompactDry™ ETB, Enterobacteriaceae, Hardy Diagnostics
  10753-986
Product available on GSA Advantage®
 :  
undefined
CompactDry™ ETB, Enterobacteriaceae, Hardy Diagnostics
  10753-986
 :  ETB100
Product available on GSA Advantage®
 :  

 

  • Description:
    CompactDry ETB Cassette
  • Size:
    100/pack
  • Storage Temperature:
    2 to 30 °C away from direct light
  • Cat. No.:
    10753-986
  • Application:
    ETB, Enterobacteriaceae
  • Supplier no.:
    ETB100

 

 

CompactDry™ is a ready-to-use system that reduces the time needed to perform microbial testing on food, cannabis, meat, cosmetics, raw materials and more.

  • Sturdy frame, stackable packaging
  • Self-diffusing media
  • Ability to subculture without damage to colonies
  • No leakage
  • Easy to label

The user friendly nature of the CompactDry™ plate allows for quick adoption in cannabis and food testing labs, regardless of technician experience.

Colonies from the Enterobacteriaceae group will turn red or purple due to the presence o+T14+K14:U14+J14:U14.

 : This product is not intended for use in the diagnosis of human disease.
 : Stored at room temperature vs the competitor products with storage at temperatures less than or equal to 8 °C.