Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Eisco® Human Testis Model

Eisco® Human Testis Model

Human testis model, 1, 13 cm, 13 cm, 24 cm
Catalog # 470136-168
Supplier:   Wards
Supplier #:  AM0115A
Eisco® Human Testis Model
Catalog # 470136-168
Supplier:  Wards
Eisco® Human Testis Model
Eisco® Human Testis Model

Human testis model, 1, 13 cm, 13 cm, 24 cm

Price
$75.65
Log in for availability
Each
Product Details & Documents

Enlarged replica a human testes with epididymis.

  • Cutaway view reveals detailed internal anatomy of this male reproductive organ
  • Designed by medical professionals and educators
  • Custom molded using the highest quality nontoxic polymers and PVC
  • Hand-painted by skilled artists
  • Includes a key card

This hand painted model of a human testis is enlarged for in-depth study. The one-piece model is rod mounted on a base with ten numbered structures identified on the included key card.

Documents

Specifications
  • Description:
    Human testis model
  • Height:
    24 cm
  • Length:
    13 cm
  • Width:
    13 cm
  • No. of parts:
    1