About this item
A premixed solution containing sodium salts of dATP, dCTP, dGTP and dTTP, each at 10mM in water.
- Ready to Use Solution of Deoxynucleotide Triphosphates
- Highly pure (>99% triphosphate)
- 10mM each nucleotide in water
- Reduce pipetting steps by adding nucleotides as a mix
dNTP Mix is a premixed solution containing sodium salts of dATP, dCTP, dGTP and dTTP, each at 10mM in water at pH 7.5; the total concentration of nucleotides is 40mM. One microliter of the dNTP Mix in a 50ul reaction will give a final dNTP concentration of 200uM for each dNTP.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Environmentally Preferable: