Specifications
- Antibody Type:Primary
- Antigen Name:6X HIS Affinity Tag
- Antigen Symbol:6xHis
- Clonality:Monoclonal
- Clone:1B7G5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:Histidine Epitope Tag|His Tag|Histidine Tag
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Protein G Purification
- Cat. No.:10082-264
- Supplier no.:66005-1-IG
Specifications
About this item
Protein tags are protein or peptide sequences located either on the C- or N- terminal of the target protein, which facilitates one or several of the following characteristics: solubility, detection, purification, localization and expression. His tag is often used for affinity purification and binding assays. Expressed His-tagged proteins can be purified and detected easily because the string of histidine residues binds to several types of immobilized metal ions, including nickel, cobalt and copper, under specific buffer conditions. The His tag antibody is a useful tool for monitoring of the His-tagged proteins, and recognizes His-tags placed at N-terminal, C-terminal, and internal regions of fusion proteins expressed in bacteria, insect, and mammalian cells.
Western Blot:recombinant Protein, 1:5000-1:50000; IP:Transfected HEK-293 Cells, 1:5000-1:50000; IF:Transfected HEK-293 Cells, 1:10-1:100
Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: 1B7G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: