Anti-transcription termination factor-Specific Mouse Monoclonal Antibody [clone: 7B3H2]
Catalog # 10081-254
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:transcription termination Factor
- Antigen Symbol:TTF1
- Clonality:Monoclonal
- Clone:7B3H2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Protein A Purification
- Cat. No.:10081-254
- Supplier no.:60235-1-IG
Specifications
About this item
TTF1 is a multifunctional nucleolar protein that terminates ribosomal gene transcription, mediates replication fork arrest and regulates RNA polymerase I transcription on chromatin. It plays a dual role in rDNA regulation, being involved in both activation and silencing of rDNA transcription. Interaction with BAZ2A/TIP5, TTF1 recovers DNA-binding activity. The antibody is specific to TTF1.
Western Blot: Hela cell, 1:1000-1:10000
Type: Primary
Antigen: transcription termination factor-Specific
Clonality: Monoclonal
Clone: 7B3H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse