Anti-MYH11 Mouse Monoclonal Antibody [clone: 5C6D3]
Catalog # 10081-642
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Myosin Heavy Chain 11
- Antigen Symbol:MYH11
- Clonality:Monoclonal
- Clone:5C6D3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:AV071570|SM2|SM1|smMHC
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Protein G Purification
- Cat. No.:10081-642
- Supplier no.:60222-1-IG
Specifications
About this item
Western Blot: Hela Cells, 1:200-1:2000
Type: Primary
Antigen: MYH11
Clonality: Monoclonal
Clone: 5C6D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse