Anti-PYCR2 Rabbit Polyclonal Antibody
10081-428
- Antibody Type:Primary
- Antigen Name:Pyrroline-5-carboxylate Reductase 2
- Antigen Symbol:PYCR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Cat. No.:10081-428
- Supplier no.:55060-1-AP
PYCR2, also named as P5CR2, belongs to the pyrroline-5-carboxylate reductase family. It catalyses the reaction: L-proline + NAD(P)+ = 1-pyrroline-5-carboxylate + NAD(P)H. The antibody is specific to PYCR2. It has no cross reaction to PYCR1.
Western Blot: HeLa Cells, 1:200-1:2000; IF: HeLa Cells, 1:20-1:200
Type: Primary
Antigen: PYCR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human