Specifications
- Antibody Type:Primary
- Antigen Name:ATPase, H+ transporting, lysosomal 70kDa, V1 Subunit A
- Antigen Symbol:ATP6V1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:ATP6V1A
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Cat. No.:10081-956
- Supplier no.:51152-1-AP
Specifications
About this item
The vacuolar-type H(+)-ATPase (V-ATPase) is responsible for the acidification of endosomes, lysosomes, and other intracellular organelles. It is also involved in hydrogen ion transport across the plasma membrane into the extracellular space. The V-ATPase is a multisubunit complex with cytosolic and transmembrane domains. The cytosolic catalytic domain consists of 3 A subunits and 3 B subunits, which bind and hydrolyze ATP, as well as regulatory accessory subunits.ATP6V1A is V-type proton ATPase catalytic subunit A.This antibody is specific to ATP6V1A.
Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Testis Tissue, 1:20-1:200
Type: Primary
Antigen: ATP6V1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse