Specifications
- Antibody Type:Primary
- Antigen Name:Heparanase
- Antigen Symbol:HPSE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:HSE1.|HPSE|HPSE 1|heparanase|HPR 1|Hpa1|Endo-glucoronidase|HEP|Hpa 1|heparanase 1|HPR1|Heparanase 50 kDa subunit|HPSE1|heparanase-1|Heparanase1|HPSE_HUMAN|HPA|Heparanase 1|heparanase1|Endo glucoronidase|HSE 1
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10081-852
- Supplier no.:24529-1-AP
Specifications
About this item
HPSE(Heparanase) is also named as HEP, HPA, HPA1, HPR1, HPSE1, HSE1 and belongs to the glycosyl hydrolase 79 family. It is a endoglycosidase that cleaves heparan sulfate proteoglycans (HSPGs) into heparan sulfate side chains and core proteoglycans. HPSE is essential in the disassembly of the extracellular matrix (ECM) by invading cells. It has 3 isoforms produced by alternative splicing with the molecular weight of 61 kDa, 55 kDa and 53 kDa. The full length protein has six glycosylation sites. The cleavage of the 65 kDa form leads to the generation of a linker peptide, and 8 kDa and 50 kDa products. The active form, the 8/50 kDa heterodimer, is resistant to degradation and glycosylation of the 50 kDa subunit appears to be essential for its solubility.
Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human Liver cancer Tissue, 1:20-1:200; IF: HeLa Cells, 1:10-1:100
Type: Primary
Antigen: HPSE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse