Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-GPR50 Rabbit Polyclonal Antibody
  10081-816
Anti-GPR50 Rabbit Polyclonal Antibody
  10081-816
 :  21514-1-AP-150UL

 

  • Antibody Type:
    Primary
  • Antigen Name:
    G Protein-coupled Receptor 50
  • Antigen Symbol:
    GPR50
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    H9|Mel1c
  • Storage Buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Fusion Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10081-816
  • Supplier no.:
    21514-1-AP

 

 

IF:SH-SY5Y Cells, N/A; Western Blot:transfected Cells, N/A

Type: Primary
Antigen: GPR50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse