Anti-CHAT Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
CHAT (choline acetyltransferase) belongs to the carnitine/choline acetyltransferase family. It catalyzes the reversible synthesis of acetylcholine (ACh) from acetyl CoA and choline at cholinergic synapses. Defects in CHAT are the cause of congenital myasthenic syndrome with episodic apnea (CMSEA). CHAT is known to to be present exclusively in the cytoplasm of cholinergic neurons and anti-CHAT antibody has been considered as the best marker for cholinergic neurons. It has been observed that CHAT gene generates multiple mRNA splice variants, including peripheral type CHAT (pCHAT) and common type CHAT (cCHAT), encoding 40-55 kDa and 66-70 kDa forms of proteins, respectively. This antibody mainly recognizes the 45-50 kDa pCHAT in multiple lysates.
Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human Stomach, 1:20-1:200; IF: HepG2, 1:10-1:100
Type: Primary
Antigen: CHAT
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Documents
Proteintech Antibodies Neuroscience
Product Literature
Specifications
- Size:150 µL
- Clonality:Polyclonal
- Purification:Antigen affinity Purification
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- ImmunoFluorescence:Yes
- Isotype:IgG
- Host:Rabbit
- Cat. No.:10082-010
- Reactivity:Human|Rat|Mouse
- Western Blot:Yes
- Shipping Temperature:Store at -20°C
- Antibody Type:Primary
- Antigen Symbol:CHAT
- Immunogen:Peptide
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:B230380D24Rik|CMS1A2|choline acetylase|choline O-acetyltransferase|CHOACTase|CMS1A
- Antigen Name:Choline Acetyltransferase
- ELISA:Yes
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



