Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-CK7 Rabbit Polyclonal Antibody
  10081-268
Anti-CK7 Rabbit Polyclonal Antibody
  10081-268
 :  17513-1-AP-150UL

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Cytokeratin 7
  • Antigen Symbol:
    CK7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    CK7|cytokeratin 7|sarcolectin|SCL|type II cytoskeletal 7|type-II keratin Kb7|K2C7|type II mesothelial keratin K7|CK-7|K7|cytokeratin-7|keratin-7|55K type II cytoskeletal|keratin
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Peptide
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10081-268
  • Supplier no.:
    17513-1-AP

 

 

Keratins are a large family of proteins that form the intermediate filament cytoskeleton of epithelial cells, which are classified into two major sequence types. Type I keratins are a group of acidic intermediate filament proteins, including K9–K23, and the hair keratins Ha1–Ha8. Type II keratins are the basic or neutral courterparts to the acidic type I keratins, including K1–K8, and the hair keratins, Hb1–Hb6. KRT7 is a type II keratin. It is specifically expressed in the simple epithelia lining the cavities of the internal organs and in the gland ducts and blood vessels. This antibody is specifically against KRT7.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: CK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: