Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-PYCR2 Rabbit Polyclonal Antibody
  10081-426
undefined
Anti-PYCR2 Rabbit Polyclonal Antibody
  10081-426
 :  17146-1-AP-150UL

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Pyrroline-5-carboxylate Reductase 2
  • Antigen Symbol:
    PYCR2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10081-426
  • Supplier no.:
    17146-1-AP

 

 

Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human ovary Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: PYCR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat