Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CHPF Rabbit Polyclonal Antibody
Anti-CHPF Rabbit Polyclonal Antibody
Catalog # 10082-020
Supplier:  Proteintech
undefined
Anti-CHPF Rabbit Polyclonal Antibody
Catalog # 10082-020
Supplier:  Proteintech
Supplier Number:  15801-1-AP-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Chondroitin Polymerizing Factor
  • Antigen Symbol:
    CHPF
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    CSS2|CHSY2
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10082-020
  • Supplier no.:
    15801-1-AP

Specifications

About this item

Western Blot: Human Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: CHPF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: