- Antibody Type:Primary
- Antigen Name:Inosine-5'-monophosphate Dehydrogenase 2
- Antigen Symbol:IMPDH2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:inosine 5' phosphate dehydrogenase 2|IMPD2|inosine monophosphate dehydrogenase type II|IMP oxireductase 2|inosine monophosphate oxireductase 2|IMPDH2|IMP dehydrogenase 2|inosine monophosphate dehydrogenase 2|IMP (inosine monophosphate) dehydrogenase 2|IMPDH-II
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10081-574
- Supplier no.:12948-1-AP
IMPDH2, also named as IMPD2, belongs to the IMPDH/GMPR family. It is a rate limiting enzyme in the de novo synthesis of guanine nucleotides and therefore is involved in the regulation of cell growth. It may also have a role in the development of malignancy and the growth progression of some tumors. IMPDH2 is a key enzyme in the purine nucleotide biosynthetic pathway and constitutes a pivotal biological target for immunosuppressant and antiviral drugs. IMPDH2 is highly enriched in RR(filamentous rods and rings).
Western Blot: Human Heart Tissue, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; IP:K-562 Cells, 1:200-1:2000
Type: Primary
Antigen: IMPDH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat