Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CaMKII gamma Rabbit Polyclonal Antibody
Anti-CaMKII gamma Rabbit Polyclonal Antibody
  10081-990
Anti-CaMKII gamma Rabbit Polyclonal Antibody
  10081-990
 :  12666-2-AP-150UL

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Calcium/Calmodulin-dependent Protein Kinase Gamma 2
  • Antigen Symbol:
    CAMK2G
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Storage Buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10081-990
  • Supplier no.:
    12666-2-AP

 

 

CaMKII gamma, also named as CAMKG, belongs to the protein kinase superfamily, CAMK Ser/Thr protein kinase family and CaMK subfamily. CAMK2 is a prominent kinase in the central nervous system that may function in long-term potentiation and neurotransmitter release. CAMK2G has been shown to be expressed in human beta cells as a candidate gene for Type II (non-insulin-dependent) diabetes mellitus. Some study can be used to assess the role of CAMK2G in the susceptibility to Type II diabetes. This antibody can recognize all the 9 isoforms of CAMK2G. So the band in WB is from 50-65kd. This antibody can bind all the CAMK2s for the close sequences.

Western Blot:SMMC-7721 Cells, 1:1000-1:10000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: CaMKII gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat