Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-PRKACB Rabbit Polyclonal Antibody
Anti-PRKACB Rabbit Polyclonal Antibody
Catalog # 10081-196
Supplier:  Proteintech
Anti-PRKACB Rabbit Polyclonal Antibody
Catalog # 10081-196
Supplier:  Proteintech
Supplier Number:  12232-1-AP-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Protein Kinase, cAMP-dependent, Catalytic Beta
  • Antigen Symbol:
    PRKACB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    PKA C-beta|cAMP-dependent protein kinase|PKACB|CbPKA|beta-catalytic subunit
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10081-196
  • Supplier no.:
    12232-1-AP

Specifications

About this item

PRKACB, also named as PKA C-beta, belongs to the protein kinase superfamily. AGC Ser/Thr protein kinase family and cAMP subfamily. It mediates cAMP-dependent signaling triggered by receptor binding to GPCRs. PKA activation regulates diverse cellular processes such as cell proliferation, the cell cycle, differentiation and regulation of microtubule dynamics, chromatin condensation and decondensation, nuclear envelope disassembly and reassembly, as well as regulation of intracellular transport mechanisms and ion flux.

Western Blot: HT-1080 Cells, 1:1000-1:10000; IHC: Human medulloblastoma Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: PRKACB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat