Anti-CAPZA1 Rabbit Polyclonal Antibody
Catalog # 10081-994
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Capping protein (actin filament) muscle Z-line, alpha 1
- Antigen Symbol:CAPZA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:CAPPA1|CAZ1|CAPZ
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10081-994
- Supplier no.:11806-1-AP
Specifications
About this item
CAPZA1 (F-actin-capping protein subunit alpha-1) is a member of the F-actin capping protein alpha subunit family and regulates actin filament growth, stability and disassembly. Differential expression of CAPZA1 has been found in gastric cancer and melanoma, indicating its possible role in tumorigenesis. (21566537, 23545944)
Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human ovary tumor Tissue, 1:20-1:200
Type: Primary
Antigen: CAPZA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat