Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-14-3-3 epsilon Rabbit Polyclonal Antibody
Anti-14-3-3 epsilon Rabbit Polyclonal Antibody
Catalog # 10082-254
Supplier:  Proteintech
undefined
Anti-14-3-3 epsilon Rabbit Polyclonal Antibody
Catalog # 10082-254
Supplier:  Proteintech
Supplier Number:  11648-2-AP-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    14-3-3 epsilon
  • Antigen Symbol:
    14-3-3 epsilon
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Storage Buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10082-254
  • Supplier no.:
    11648-2-AP

Specifications

About this item

14-3-3 epsilon (also known as YWHAE) is a member of 14-3-3 proteins which were the first phosphoserine/phosphothreonine-binding proteins to be discovered. 14-3-3 family members interact with a wide spectrum of proteins and possess diverse functions. Mammals express seven distinct 14-3-3 isoforms (gamma, epsilon, beta, zeta, sigma, theta, tau) that form multiple homo- and hetero- dimmers. 14-3-3 proteins display the highest expression levels in the brain, and have been implicated in several neurodegenerative diseases, including Alzheimer's disease and amyotrophic lateral sclerosis. This antibody was raised against full-length 14-3-3 epsilon.

Western Blot:A375 Cells, 1:1000-1:10000; IHC: Human gliomas Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP:A375 Cells, 1:500-1:5000; FC: HepG2 Cells, N/A

Type: Primary
Antigen: 14-3-3 epsilon
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat