Specifications
- Antibody Type:Primary
- Antigen Name:Tubulin alpha 1
- Antigen Symbol:TUBA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:tubulin alpha-4A chain|alpha 1|tubulin H2-alpha|tubulin|tubulin alpha-1 chain|H2-ALPHA
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10081-874
- Supplier no.:11224-1-AP
Specifications
About this item
There are five tubulins in human cells: alpha, beta, gamma, delta, and epsilon. Tubulins are conserved across species. They form heterodimers, which multimerize to form a microtubule filament. An alpha and beta tubulin heterodimer is the basic structural unit of microtubules. The heterodimer does not come apart, once formed. The alpha and beta tubulins, which are each about 55 kDa MW, are homologous but not identical. Alpha, beta, and gamma tubulins have all been used as loading controls. Tubulin expression may vary according to resistance to antimicrobial and antimitotic drugs.
IHC: Human Colon cancer Tissue, 1:20-1:200; Western Blot: Multi-Cells/Tissue, 1:500-1:5000; IP: HeLa Cells, 1:500-1:5000; IF: MCF-7 Cells, 1:10-1:100; FC: HEK-293 Cells, N/A
Type: Primary
Antigen: TUBA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat