Chromatography Basic Glass Columns with PTFE Stopcock Plug, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
A general purpose column for analytical separations.
A plug of glass wool can be used to support the column packing.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Frequently Bought Together