Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Hamster IgG Rabbit Polyclonal Antibody (Biotin (NHS-LC))
Anti-Hamster IgG Rabbit Polyclonal Antibody (Biotin (NHS-LC))
Catalog # 10151-198
Supplier:  Immunoreagents
undefined
Anti-Hamster IgG Rabbit Polyclonal Antibody (Biotin (NHS-LC))
Catalog # 10151-198
Supplier:  Immunoreagents
Supplier Number:  RBXHA-003-EBIO

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin (NHS-LC)
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Hamster
  • Western Blot:
    Yes
  • Size:
    1.0 mg
  • Cross Adsorption:
    Yes
  • Format:
    Purified IgG
  • Form:
    Clear, colorless liquid
  • Storage Buffer:
    10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
  • Storage Temperature:
    2-8 °C
  • Concentration:
    1.50 mg/ml (E 1% at 280 nm = 13.0)
  • Shipping Temperature:
    2-8 °C
  • Immunogen:
    Hamster IgG
  • Tested Applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Affinity purified using solid phaseHamster IgG
  • Cat. No.:
    10151-198
  • Supplier no.:
    IR2560

Specifications

About this item

ImmunoReagents’ Biotinylated antibody conjugates are widely used due to their ability to enhance antibody signal in most immunoassays. Biotinylated secondary antibodies are used in most immunoassay formats (i.e., ELISA, Immunohistochemistry (IHC), and Western blotting). While Biotinylated antibodies by themselves do not produce a signal that can be visualized, biotin has a high affinity and specificity to Streptavidin/Avidin. Using ImmunoReagents’ streptavidin conjugates with biotinylated antibodies results in signal amplification.

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin (NHS-LC)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster