Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-IgE epsilon Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-IgE epsilon Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  10149-724
undefined
Anti-IgE epsilon Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  10149-724
 :  GTXHU-002-DFITC

 

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin E epsilon
  • Antigen Symbol:
    IgE
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgE
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    1.0 mg
  • Cross Adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Clear, fluorescent yellow liquid
  • Storage Buffer:
    10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
  • Storage Temperature:
    2-8 °C
  • Concentration:
    1.50 mg/ml (E 1% at 280 nm = 13.0)
  • Shipping Temperature:
    2-8 °C
  • Immunogen:
    Human IgE, (µ) chain
  • Tested Applications:
    Fluoresence ELISA, Fluorescence-Activated Cell Sorting
  • Purification:
    Affinity purified using solid phase Human IgE
  • Cat. No.:
    10149-724
  • Supplier no.:
    IR1561

 

 

ImmunoReagents’ FITC Conjugates are used in immunofluorescence microscopy, flow cytometry, FLISA (fluorescent ELISA) and high throughput screening assays. Fluorescein is typically excited by the 488 nm line of an argon laser, and emission is collected at 518 nm. FITC antibody conjugates are yellow-orange in color but emit a yellow-green color upon Em 518 nm. FITC antibody conjugates are sensitive to light. The typical FITC to antibody ratio (F/P) is >4.0 Excitation/Emission = 494nm / 518nm Emission Color = Green (Similar Dyes: Alexa Fluor 488, Cy2, DyLight ® 488)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

 : (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
 : Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgE epsilon
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgE
Reactivity: Human