Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-IgE epsilon Goat Polyclonal Antibody (Biotin (NHS-LC))
  10151-908
undefined
Anti-IgE epsilon Goat Polyclonal Antibody (Biotin (NHS-LC))
  10151-908
 :  GTXHU-002-DBIO

 

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin E epsilon
  • Antigen Symbol:
    IgE
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin (NHS-LC)
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgE
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    1.0 mg
  • Cross Adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Clear, colorless liquid
  • Storage Buffer:
    10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
  • Storage Temperature:
    2-8 °C
  • Concentration:
    1.50 mg/ml (E 1% at 280 nm = 13.0)
  • Shipping Temperature:
    2-8 °C
  • Immunogen:
    Human IgE, (µ) chain
  • Tested Applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Affinity purified using solid phase Human IgE
  • Cat. No.:
    10151-908
  • Supplier no.:
    IR1547

 

 

ImmunoReagents’ Biotinylated antibody conjugates are widely used due to their ability to enhance antibody signal in most immunoassays. Biotinylated secondary antibodies are used in most immunoassay formats (i.e., ELISA, Immunohistochemistry (IHC), and Western blotting). While Biotinylated antibodies by themselves do not produce a signal that can be visualized, biotin has a high affinity and specificity to Streptavidin/Avidin. Using ImmunoReagents’ streptavidin conjugates with biotinylated antibodies results in signal amplification.

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

 : (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
 : Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgE epsilon
Clonality: Polyclonal
Clone:
Conjugation: Biotin (NHS-LC)
Epitope:
Host: Goat
Isotype: IgE
Reactivity: Human