Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-IgY Goat Polyclonal Antibody (DyLight® 350)
  10149-614
undefined
Anti-IgY Goat Polyclonal Antibody (DyLight® 350)
  10149-614
 :  GTXCK-003-D350NHSX

 

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin Y
  • Antigen Symbol:
    IgY
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 350
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgY
  • Reactivity:
    Chicken
  • Western Blot:
    Yes
  • Size:
    1.0 mg
  • Cross Adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Lyophilized
  • Storage Buffer:
    10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
  • Storage Temperature:
    2-8 °C
  • Concentration:
    1 mg/ml (E 1% at 280 nm = 13.0)
  • Shipping Temperature:
    2-8 °C
  • Immunogen:
    Chicken IgG/Y
  • Tested Applications:
    Fluoresence ELISA, Fluorescence-Activated Cell Sorting
  • Purification:
    Affinity purified using solid phaseChicken IgG/Y
  • Cat. No.:
    10149-614
  • Supplier no.:
    IR1410

 

 

The ImmunoReagents' DyLight® 350 conjugates are a derivative of AMCA (Aminomethylcoumarine acetate), which a blue fluorescent dye commonly is used as a contrasting dye for double- and triple-labeling used in a wide array of applications. The UV-excitable DyLight® 350 has spectral properties that are very similar to those displayed by AMCA, including identical excitation wavelengths and a large stokes shift. The emission wavelength of DyLight® 350 has a 10-nm blue shift over AMCA, though, that provides an advantage in multiplexing with green fluorophores. (Excitation/Emission = 353nm / 432nm Emission Color = Blue (Similar Dyes: Alexa Fluor 350, AMCA)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

 : (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
 : Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 350
Epitope:
Host: Goat
Isotype: IgY
Reactivity: Chicken