Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-AFP Goat Polyclonal Antibody
Anti-AFP Goat Polyclonal Antibody
Catalog # 10149-534
Supplier:  Immunoreagents
undefined
Anti-AFP Goat Polyclonal Antibody
Catalog # 10149-534
Supplier:  Immunoreagents
Supplier Number:  GTXHU-055-B1

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Alpha 1 Fetoprotein Receptor
  • Antigen Symbol:
    AFP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Western Blot:
    Yes
  • Size:
    1.0 ml
  • Cross Adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Clear, colorless liquid, 0.2 µm filtered
  • Antigen Synonyms:
    FETA|Alpha-1-Fetoprotein|HPAFP
  • Storage Buffer:
    10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2
  • Storage Temperature:
    2-8 °C
  • Concentration:
    >10 mg/ml (E 1% at 280 nm = 1.3)
  • Shipping Temperature:
    2-8 °C
  • Immunogen:
    Human Alphafetoprotein (AFP)
  • Tested Applications:
    Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
  • Purification:
    Proprietary 2-step fractionation procedure
  • Cat. No.:
    10149-534
  • Supplier no.:
    IR1366

Specifications

About this item

ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.  

  • Origin: US

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Type: Primary
Antigen: AFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human